Assay Details
Target Gene Details
Entrez Gene ID: | 72789 |
Gene Name: | ventricular zone expressed PH domain-containing 1 |
Gene Aliases: |
2810471M23Rik, Veph |
Location: |
Chr.3:66053558-66297066 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Veph1 | NM_145820.3 | NP_665819.2 | ||
| AB085187.1 | BAC02920.1 | |||
| AB085189.1 | BAC02922.1 | |||
| AK013398.1 | BAB28831.1 | |||
| BC128271.1 |

Genomic Map