Assay Details
Target Gene Details
Entrez Gene ID: | 54325 |
Gene Name: | elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 1 |
Gene Aliases: |
AA407424, BB151133, Ssc1 |
Location: |
Chr.4:118428081-118432953 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elovl1 | NM_001039175.2 | NP_001034264.1 | ||
| NM_001039176.2 | NP_001034265.1 | |||
| NM_019422.3 | NP_062295.1 | |||
| AF170907.1 | AAF72572.1 | |||
| AK003743.1 | BAB22975.1 | |||
| AK135348.1 | BAE22497.1 | |||
| AK156003.1 | 1 | 1614 | BAE33541.1 | |
| AK171217.1 | BAE42321.1 | |||
| BC006735.1 | AAH06735.1 | |||
| BC096673.1 | AAH96673.1 | |||
| BY141978.1 | ||||
| BY773043.1 | ||||
| CJ094566.1 | ||||
| CX199666.1 |
More Information
Set Membership: |
|

Genomic Map