Assay Details
Target Gene Details
Entrez Gene ID: | 14579 |
Gene Name: | GTP binding protein (gene overexpressed in skeletal muscle) |
Gene Aliases: |
AV020497 |
Location: |
Chr.4:11704447-11714993 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gem | NM_010276.4 | NP_034406.2 | ||
| AK157655.1 | ||||
| AK158393.1 | BAE34484.1 | |||
| AK168401.1 | BAE40322.1 | |||
| AK171336.1 | BAE42399.1 | |||
| BC029668.1 | AAH29668.1 | |||
| U10551.1 | AAA64912.1 | |||
| U13053.1 | AAC52145.1 |

Genomic Map