Assay Details
Target Gene Details
Entrez Gene ID: | 67694 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
1700029H06Rik, Ccdc2, Cmg1, b2b796Clo |
Location: |
Chr.4:94603266-94693233 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift74 | NM_001290568.1 | 9 | 820 | NP_001277497.1 |
| NM_026319.3 | 9 | 820 | NP_080595.2 | |
| AK011690.1 | 9 | 791 | BAB27780.1 | |
| AK014770.1 | 9 | 788 | BAB29542.3 | |
| AK053369.1 | 9 | 768 | BAC35365.1 | |
| BC005798.1 | 9 | 790 | ||
| BC023760.1 | 9 | 752 | AAH23760.1 | |
| BQ950148.1 | 9 | 792 |

Genomic Map