Assay Details
Target Gene Details
Entrez Gene ID: | 14201 |
Gene Name: | four and a half LIM domains 3 |
Gene Aliases: |
SLIM2 |
Location: |
Chr.4:124695963-124708611 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fhl3 | NM_010213.3 | 3 | 452 | NP_034343.2 |
| AF114382.1 | 3 | 423 | AAD53231.1 | |
| AF134772.1 | 3 | 449 | AAD32623.1 | |
| AF149826.1 | 2 | 250 | AAF73159.1 | |
| BC144903.1 | 3 | 416 | ||
| BC145939.1 | 3 | 416 | ||
| CB205184.1 | 1 | 109 |
More Information
Set Membership: |
|

Genomic Map