Assay Details
Target Gene Details
Entrez Gene ID: | 14700 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 10 |
Gene Aliases: |
AV006524 |
Location: |
Chr.4:59035156-59041899 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gng10 | NM_025277.3 | NP_079553.1 | ||
| AK003187.1 | ||||
| AK014158.1 | BAB29183.1 | |||
| BC023670.1 | AAH23670.1 | |||
| BC055434.1 | AAH55434.1 | |||
| BY274678.1 |
More Information
Set Membership: |
|

Genomic Map