Assay Details
Target Gene Details
Entrez Gene ID: | 194231 |
Gene Name: | connector enhancer of kinase suppressor of Ras 1 |
Gene Aliases: |
BC037758 |
Location: |
Chr.4:134228041-134238416 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnksr1 | NM_001081047.1 | 20 | 1872 | NP_001074516.1 |
| AK145910.1 | 20 | 1834 | ||
| BC113805.1 | 20 | 1837 | ||
| BC113956.1 | 20 | 1837 |

Genomic Map