Assay Details
Target Gene Details
Entrez Gene ID: | 15502 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 1 |
Gene Aliases: |
Hsj2, Nedd7 |
Location: |
Chr.4:40722468-40737153 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja1 | NM_008298.5 | 1 | 31 | NP_032324.1 |
| AF055664.1 | AAC78597.1 | |||
| AK049963.1 | ||||
| AK083046.1 | BAC38744.1 | |||
| AK145963.1 | BAE26788.1 | |||
| AK167139.1 | BAE39284.1 | |||
| AK167212.1 | BAE39339.1 | |||
| BB574002.1 | 1 | 33 |
Target Gene Details
Entrez Gene ID: | 622208 |
Gene Name: | predicted gene 6297 |
Gene Aliases: |
- |
Location: |
Chr.4:40720154-40722841 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm6297 | NR_077226.1 | |||
| AK139286.1 | 1 | 370 | ||
| BE656271.1 |
More Information
Set Membership: |
|

Genomic Map