Assay Details
Target Gene Details
Entrez Gene ID: | 242297 |
Gene Name: | family with sequence similarity 110, member B |
Gene Aliases: |
1700012H17Rik, 5031405P22, AW049855 |
Location: |
Chr.4:5644078-5801101 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam110b | NM_173426.2 | NP_775602.1 | ||
| AK052596.1 | BAC35053.1 | |||
| AK080588.1 | ||||
| AK159054.1 | BAE34787.1 | |||
| BC055683.1 | AAH55683.1 |

Genomic Map