Assay Details
Target Gene Details
Entrez Gene ID: | 277743 |
Gene Name: | family with sequence similarity 131, member C |
Gene Aliases: |
Gm693 |
Location: |
Chr.4:141368192-141384174 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam131c | NM_001085513.2 | 2 | 199 | NP_001078982.1 |
| BC147048.1 | 2 | 199 | ||
| BC147049.1 | 2 | 199 |

Genomic Map