Assay Details
Target Gene Details
Entrez Gene ID: | 67389 |
Gene Name: | family with sequence similarity 132, member A |
Gene Aliases: |
1110035L05Rik, C1qdc2, CTRP12, alipolin |
Location: |
Chr.4:155962312-155966629 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam132a | NM_026125.3 | NP_080401.1 | ||
| AK002876.1 | BAB22423.1 | |||
| AK003510.1 | BAB22827.1 | |||
| AK004116.1 | BAB23178.1 | |||
| AK007335.1 | ||||
| BC026939.1 | AAH26939.1 | |||
| DQ002404.1 | AAY21936.1 |
More Information
Set Membership: |
|

Genomic Map