Assay Details
Target Gene Details
Entrez Gene ID: | 338355 |
Gene Name: | FK506 binding protein 15 |
Gene Aliases: |
BB131447, C430014M02Rik, FKBO133, FKBP-133, FKBP-15, FKBP133, mKIAA0674 |
Location: |
Chr.4:62300342-62360591 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp15 | NM_001045528.1 | 22 | 2428 | NP_001038993.1 |
| AK122343.1 | 15 | 1667 | BAC65625.1 | |
| AK170070.1 | 2 | 173 | ||
| BC060651.1 | 22 | 2342 | AAH60651.1 | |
| BC137706.1 | 22 | 2332 |

Genomic Map