Assay Details
Target Gene Details
Entrez Gene ID: | 15502 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily A, member 1 |
Gene Aliases: |
Hsj2, Nedd7 |
Location: |
Chr.4:40722468-40737153 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaja1 | NM_001164671.1 | NP_001158143.1 | ||
| NM_001164672.1 | NP_001158144.1 | |||
| NM_008298.5 | NP_032324.1 | |||
| AF055664.1 | AAC78597.1 | |||
| AK049963.1 | ||||
| AK083046.1 | BAC38744.1 | |||
| AK145963.1 | BAE26788.1 | |||
| AK147604.1 | ||||
| AK167139.1 | BAE39284.1 | |||
| AK167212.1 | BAE39339.1 | |||
| BC057876.1 | AAH57876.1 | |||
| BC158024.1 |
More Information
Set Membership: |
|

Genomic Map