Assay Details
Target Gene Details
Entrez Gene ID: | 338355 |
Gene Name: | FK506 binding protein 15 |
Gene Aliases: |
BB131447, C430014M02Rik, FKBO133, FKBP-133, FKBP-15, FKBP133, mKIAA0674 |
Location: |
Chr.4:62300342-62360591 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp15 | NM_001045528.1 | 9 | 954 | NP_001038993.1 |
| AK049459.1 | 9 | 899 | ||
| AK049509.1 | 9 | 955 | ||
| AK082565.1 | 9 | 903 | ||
| AK085087.1 | 9 | 942 | ||
| AK122343.1 | 2 | 193 | BAC65625.1 | |
| AK161705.1 | 9 | 884 | BAE36541.1 | |
| BC060651.1 | 9 | 868 | AAH60651.1 | |
| BC137706.1 | 9 | 888 |

Genomic Map