Assay Details
Target Gene Details
Entrez Gene ID: | 15444 |
Gene Name: | hippocalcin |
Gene Aliases: |
- |
Location: |
Chr.4:129111570-129122070 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hpca | NM_001130419.2 | NP_001123891.1 | ||
| NM_001286081.1 | NP_001273010.1 | |||
| NM_010471.4 | NP_034601.1 | |||
| AK002992.1 | BAB22502.1 | |||
| BC049607.1 | AAH49607.1 | |||
| BY128037.1 | ||||
| BY129411.1 | ||||
| BY769297.1 | ||||
| CX238650.1 | ||||
| CX239660.1 | ||||
| CX240757.1 |

Genomic Map