Assay Details
Target Gene Details
Entrez Gene ID: | 15444 |
Gene Name: | hippocalcin |
Gene Aliases: |
- |
Location: |
Chr.4:129111570-129122070 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hpca | NM_001130419.2 | NP_001123891.1 | ||
| NM_001286081.1 | NP_001273010.1 | |||
| NM_001286083.1 | NP_001273012.1 | |||
| NM_010471.4 | NP_034601.1 | |||
| AF326551.1 | AAL37397.1 | |||
| AF326552.1 | AAL37398.1 | |||
| AK002992.1 | BAB22502.1 | |||
| BC049607.1 | AAH49607.1 | |||
| BC058588.1 | AAH58588.1 | |||
| CX238650.1 | ||||
| CX239660.1 | ||||
| CX240088.1 | ||||
| CX240757.1 |

Genomic Map