Assay Details
Target Gene Details
Entrez Gene ID: | 329831 |
Gene Name: | family with sequence similarity 166, member B |
Gene Aliases: |
4833436C18Rik |
Location: |
Chr.4:43426675-43431593 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam166b | NM_001162381.1 | NP_001155853.1 | ||
| NM_177377.5 | NP_796351.2 | |||
| AK029435.1 | BAC26451.1 | |||
| AK053208.1 | BAC35311.1 |

Genomic Map