Assay Details
Target Gene Details
Entrez Gene ID: | 242546 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 12 |
Gene Aliases: |
BB197342, Cyp2j12-ps, OTTMUSG00000007939 |
Location: |
Chr.4:96099314-96141272 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j12 | NM_001100182.2 | NP_001093652.1 | ||
| AK136922.1 |
More Information
Set Membership: |
|

Genomic Map