Assay Details
Target Gene Details
Entrez Gene ID: | 230459 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 13 |
Gene Aliases: |
- |
Location: |
Chr.4:96042660-96077566 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j13 | NM_145548.4 | NP_663523.2 | ||
| AK143903.1 | BAE25592.1 | |||
| AK143981.1 | BAE25643.1 | |||
| AK165405.1 | BAE38165.1 | |||
| AK165469.1 | BAE38206.1 | |||
| AK165473.1 | BAE38210.1 | |||
| BC016446.1 | AAH16446.1 |
More Information
Set Membership: |
|

Genomic Map