Assay Details
Target Gene Details
Entrez Gene ID: | 72429 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 25 |
Gene Aliases: |
2010109C08Rik, 2010203O07Rik |
Location: |
Chr.4:58994810-59023398 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc25 | NM_001033165.3 | NP_001028337.2 | ||
| AK008359.1 | ||||
| AK008425.1 | ||||
| AK136359.1 | BAE22945.1 | |||
| BC119039.1 | ||||
| BC120693.1 |
More Information
Set Membership: |
|

Genomic Map