Assay Details
Target Gene Details
Entrez Gene ID: | 269587 |
Gene Name: | erythrocyte protein band 4.1 |
Gene Aliases: |
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056 |
Location: |
Chr.4:131923413-132075579 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1 | NM_001128606.1 | 14 | 2291 | NP_001122078.1 |
| NM_001128607.1 | 14 | 2201 | NP_001122079.1 | |
| NM_183428.3 | 17 | 3715 | NP_906273.3 | |
| AB370255.1 | 3 | 615 | ||
| AB370256.1 | 3 | 216 | ||
| AB370261.1 | 14 | 2133 | ||
| AK019923.1 | 1 | 1977 | ||
| AK133437.1 | 4 | 486 | BAE21657.1 | |
| AK147339.1 | 1 | 3078 | ||
| AK148569.1 | 1 | 985 | ||
| AK156891.1 | BAE33886.1 | |||
| AK220462.1 | 14 | 2198 | BAD90280.1 | |
| BC068138.1 | 14 | 2161 | AAH68138.1 | |
| BC079875.1 | 14 | 2120 | AAH79875.1 | |
| CB526050.1 | 7 | 658 | ||
| L00919.1 | 18 | 3790 | AAA37122.1 | |
| LK021332.1 | 14 | 1959 | ||
| LK021333.1 | 13 | 1509 |

Genomic Map