Assay Details
Target Gene Details
Entrez Gene ID: | 269587 |
Gene Name: | erythrocyte protein band 4.1 |
Gene Aliases: |
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056 |
Location: |
Chr.4:131923413-132075579 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1 | NM_001128606.1 | NP_001122078.1 | ||
| NM_001128607.1 | NP_001122079.1 | |||
| NM_183428.3 | NP_906273.3 | |||
| AB370255.1 | ||||
| AB370256.1 | ||||
| AB370261.1 | ||||
| AK133437.1 | BAE21657.1 | |||
| AK147339.1 | ||||
| AK156891.1 | BAE33886.1 | |||
| AK220462.1 | BAD90280.1 | |||
| BC068138.1 | AAH68138.1 | |||
| BC079875.1 | AAH79875.1 | |||
| L00919.1 | AAA37122.1 | |||
| LK021332.1 | ||||
| LK021333.1 |

Genomic Map