Assay Details
Target Gene Details
Entrez Gene ID: | 269587 |
Gene Name: | erythrocyte protein band 4.1 |
Gene Aliases: |
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056 |
Location: |
Chr.4:131923413-132075579 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epb4.1 | NM_001128606.1 | 9 | 1471 | NP_001122078.1 |
| NM_001128607.1 | 9 | 1381 | NP_001122079.1 | |
| NM_183428.3 | 9 | 2733 | NP_906273.3 | |
| AB370261.1 | 8 | 1256 | ||
| AK147266.1 | 4 | 427 | BAE27807.1 | |
| AK147644.1 | 4 | 427 | BAE28045.1 | |
| AK212118.1 | ||||
| AK220462.1 | 9 | 1378 | BAD90280.1 | |
| BC068138.1 | 9 | 1341 | AAH68138.1 | |
| BC079875.1 | 9 | 1300 | AAH79875.1 | |
| CN526385.1 | 3 | 315 | ||
| L00919.1 | 10 | 2808 | AAA37122.1 | |
| LK021332.1 | 7 | 626 | ||
| LK021333.1 | 7 | 626 |

Genomic Map