Assay Details
Target Gene Details
Entrez Gene ID: | 13109 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 5 |
Gene Aliases: |
- |
Location: |
Chr.4:96627432-96664170 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j5 | NM_010007.4 | NP_034137.1 | ||
| AK098119.1 | ||||
| AK143932.1 | BAE25615.1 | |||
| AK149552.1 | BAE28954.1 | |||
| AK165404.1 | BAE38164.1 | |||
| BC021624.1 | AAH21624.1 | |||
| U62294.1 | AAB87635.1 |
More Information
Set Membership: |
|

Genomic Map