Assay Details
Target Gene Details
Entrez Gene ID: | 13110 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 6 |
Gene Aliases: |
- |
Location: |
Chr.4:96516138-96560835 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j6 | NM_010008.4 | NP_034138.3 | ||
| AK033614.1 | BAC28391.1 | |||
| AK045421.1 | BAC32356.1 | |||
| AK078893.1 | ||||
| AK084025.1 | ||||
| AK165663.1 | BAE38325.1 | |||
| BC050832.1 | AAH50832.1 | |||
| U62295.1 | AAB87636.1 |
More Information
Set Membership: |
|

Genomic Map