Assay Details
Target Gene Details
Entrez Gene ID: | 100038514 |
Gene Name: | predicted gene 11837 |
Gene Aliases: |
OTTMUSG00000004551 |
Location: |
Chr.4:14929829-14953031 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm11837 | NM_001243100.1 | 2 | 175 | NP_001230029.1 |
