Assay Details
Target Gene Details
Entrez Gene ID: | 53598 |
Gene Name: | dynactin 3 |
Gene Aliases: |
p24 |
Location: |
Chr.4:41714798-41723163 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dctn3 | NM_001159565.1 | NP_001153037.1 | ||
| NM_016890.4 | NP_058586.3 | |||
| AF098508.1 | AAD16226.1 | |||
| AK002720.1 | BAB22307.1 | |||
| AK003255.1 | BAB22671.1 | |||
| AK011840.1 | BAB27872.1 | |||
| AK159344.1 | BAE35006.1 | |||
| AV493721.1 | ||||
| BC061120.1 | AAH61120.1 | |||
| BY141790.1 |
More Information
Set Membership: |
|

Genomic Map