Assay Details
Target Gene Details
Entrez Gene ID: | 68598 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 8 |
Gene Aliases: |
1110021D09Rik, 2010009J04Rik, AL024084, AU019262, AU044514 |
Location: |
Chr.4:132535548-132553742 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc8 | NM_172400.3 | NP_765988.2 | ||
| AK003892.2 | ||||
| AK049526.1 | ||||
| AK088325.1 | BAC40283.1 | |||
| AK150933.1 | ||||
| AK168645.1 | ||||
| BC057191.1 | AAH57191.1 | |||
| BC066222.1 | ||||
| BC137735.1 | ||||
| BY754536.1 |

Genomic Map