Assay Details
Target Gene Details
Entrez Gene ID: | 665095 |
Gene Name: | cytochrome P450, family 2, subfamily j, polypeptide 8 |
Gene Aliases: |
Cyp2j8-ps, OTTMUSG00000007938 |
Location: |
Chr.4:96444283-96507386 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp2j8 | NM_001104927.1 | NP_001098397.1 |
More Information
Set Membership: |
|

Genomic Map