Assay Details
Target Gene Details
Entrez Gene ID: | 14105 |
Gene Name: | serine/arginine-rich splicing factor 10 |
Gene Aliases: |
FUSIP2, Fusip1, NSSR1, NSSR2, Nssr, SRrp40, Sfrs13a, Srsf13a, TASR, TASR1, TASR2 |
Location: |
Chr.4:135855728-135869908 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srsf10 | NM_001080387.2 | NP_001073856.1 | ||
| NM_001284195.1 | NP_001271124.1 | |||
| NM_001284196.1 | NP_001271125.1 | |||
| NM_010178.3 | NP_034308.1 | |||
| AB015894.1 | BAA35092.1 | |||
| AB015895.1 | BAA35093.1 | |||
| AF042383.1 | AAC70916.1 | |||
| AF060490.1 | AAC26715.1 | |||
| AK014345.1 | BAB29286.1 | |||
| AK134076.1 | ||||
| AK134289.1 | BAE22085.1 | |||
| AK135316.1 | BAE22485.1 | |||
| AK149716.1 | BAE29044.1 | |||
| AK150072.1 | BAE29284.1 | |||
| AK150734.1 | BAE29809.1 | |||
| AK151567.1 | BAE30509.1 | |||
| AK161097.1 | 2 | 3667 | ||
| AK168524.1 | BAE40403.1 | |||
| AK168558.1 | BAE40431.1 | |||
| AK168571.1 | BAE40441.1 | |||
| AK169071.1 | BAE40858.1 | |||
| BC037591.1 | AAH37591.1 | |||
| BC043060.1 | AAH43060.1 | |||
| BC083082.1 | AAH83082.1 | |||
| BY244789.1 | ||||
| BY717856.1 | ||||
| CF914989.1 | ||||
| CN528501.1 |
More Information
Set Membership: |
|

Genomic Map