Assay Details
Target Gene Details
Entrez Gene ID: | 319885 |
Gene Name: | zinc finger, CCHC domain containing 7 |
Gene Aliases: |
4930572I07Rik, D4Wsu132e |
Location: |
Chr.4:44756559-44932215 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zcchc7 | NM_138590.4 | NP_613056.2 | ||
| AK029808.1 | ||||
| AK032353.1 | ||||
| AK039158.1 | ||||
| AK043238.1 | BAC31499.1 | |||
| AK080785.1 | ||||
| AK157728.1 | BAE34173.1 | |||
| BC117020.1 | ||||
| BC117022.1 | ||||
| BC144774.1 | ||||
| CJ150974.1 | ||||
| CJ155194.1 |
More Information
Set Membership: |
|

Genomic Map