Assay Details
Target Gene Details
Entrez Gene ID: | 435815 |
Gene Name: | predicted gene 13177 |
Gene Aliases: |
OTTMUSG00000010747 |
Location: |
Chr.4:144613707-144623398 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm13177 | NM_001081248.1 | NP_001074717.1 | ||
| BC150982.1 |
More Information
Set Membership: |
|

Genomic Map