Assay Details
Target Gene Details
Entrez Gene ID: | 329993 |
Gene Name: | predicted gene 438 |
Gene Aliases: |
- |
Location: |
Chr.4:144777204-144786583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm438 | NM_001126316.1 | NP_001119788.1 | ||
| BB630382.1 |
More Information
Set Membership: |
|

Genomic Map