Assay Details
Target Gene Details
Entrez Gene ID: | 12896 |
Gene Name: | carnitine palmitoyltransferase 2 |
Gene Aliases: |
AI323697, CPTII |
Location: |
Chr.4:107903982-107923589 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cpt2 | NM_009949.2 | NP_034079.2 | ||
| AK018752.1 | BAB31387.1 | |||
| AK144441.1 | BAE25892.1 | |||
| AK150163.1 | BAE29353.1 | |||
| AK169038.1 | BAE40828.1 | |||
| BC138514.1 | ||||
| BC145859.1 | ||||
| CD545813.1 | ||||
| U01163.1 | AAA18921.1 |
More Information
Set Membership: |
|

Genomic Map