Assay Details
Target Gene Details
Entrez Gene ID: | 230777 |
Gene Name: | hypocretin (orexin) receptor 1 |
Gene Aliases: |
Ox1r, ox-1-R, ox1-R |
Location: |
Chr.4:130130217-130139691 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcrtr1 | NM_001163027.1 | 5 | 873 | NP_001156499.1 |
| NM_198959.2 | 5 | 899 | NP_945197.2 | |
| AF394596.1 | 1 | 189 | AAK71326.1 | |
| AK140137.1 | 5 | 873 | BAE24252.1 | |
| AY255599.1 | 1 | 167 | AAO85111.1 | |
| AY336083.1 | 5 | 899 | AAR01326.1 | |
| BC119582.2 | 5 | 771 | ||
| BC119583.2 | 4 | 755 |
More Information
Set Membership: |
|

Genomic Map