Assay Details
Target Gene Details
Entrez Gene ID: | 329877 |
Gene Name: | DENN/MADD domain containing 4C |
Gene Aliases: |
1700065A05Rik, AA420392 |
Location: |
Chr.4:86748555-86850603 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd4c | NM_184088.1 | 13 | 2253 | NP_908976.1 |
| AK155220.1 | 13 | 2254 | BAE33131.1 | |
| AK158927.1 | 3 | 1699 | ||
| BC146602.1 | 12 | 1878 |

Genomic Map