Assay Details
Target Gene Details
Entrez Gene ID: | 230639 |
Gene Name: | cytochrome P450, family 4, subfamily a, polypeptide 29 |
Gene Aliases: |
Cyp4a29-ps, OTTMUSG00000008612 |
Location: |
Chr.4:115242084-115254557 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4a29 | NM_001100183.1 | NP_001093653.1 |

Genomic Map