Assay Details
Target Gene Details
Entrez Gene ID: | 15569 |
Gene Name: | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) |
Gene Aliases: |
Hub, mel-N1 |
Location: |
Chr.4:91250766-91400064 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elavl2 | NM_207685.1 | NP_997568.1 | ||
| BC049125.1 | AAH49125.1 | |||
| BC058393.1 | AAH58393.1 |
More Information
Set Membership: |
|

Genomic Map