Assay Details
Target Gene Details
Entrez Gene ID: | 435802 |
Gene Name: | cytochrome P450, family 4, subfamily a, polypeptide 30b |
Gene Aliases: |
Cyp4a30b-ps, OTTMUSG00000008626 |
Location: |
Chr.4:115452491-115471418 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4a30b | NM_001100185.1 | NP_001093655.1 |

Genomic Map