Assay Details
Target Gene Details
Entrez Gene ID: | 15569 |
Gene Name: | ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) |
Gene Aliases: |
Hub, mel-N1 |
Location: |
Chr.4:91250766-91400064 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elavl2 | NM_001177883.1 | 2 | 604 | NP_001171354.1 |
| NM_010486.2 | 2 | 199 | NP_034616.1 | |
| NM_207685.1 | 3 | 311 | NP_997568.1 | |
| NM_207686.1 | 2 | 199 | NP_997569.1 | |
| AK141919.1 | 2 | 480 | BAE24886.1 | |
| AK149201.1 | 2 | 416 | ||
| AY035378.1 | 1 | 48 | AAK74152.1 | |
| AY035379.1 | 1 | 48 | AAK74153.1 | |
| AY035380.1 | 1 | 48 | AAK74154.1 | |
| BC046598.1 | 2 | 123 | AAH46598.2 | |
| BC049125.1 | 3 | 311 | AAH49125.1 | |
| BC058393.1 | 3 | 311 | AAH58393.1 | |
| BY129136.1 | ||||
| U29088.1 | 2 | 198 | AAC52644.1 |
More Information
Set Membership: |
|

Genomic Map