Assay Details
Target Gene Details
Entrez Gene ID: | 67694 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
1700029H06Rik, Ccdc2, Cmg1, b2b796Clo |
Location: |
Chr.4:94603266-94693233 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift74 | NM_001290568.1 | 11 | 1035 | NP_001277497.1 |
| NM_026319.3 | 11 | 1035 | NP_080595.2 | |
| AK006492.1 | 2 | 142 | BAB24617.1 | |
| AK011690.1 | 11 | 1006 | BAB27780.1 | |
| AK014770.1 | 11 | 1003 | BAB29542.3 | |
| AK053369.1 | 11 | 983 | BAC35365.1 | |
| BC005798.1 | 11 | 1005 | ||
| BC023760.1 | 11 | 967 | AAH23760.1 | |
| BY706530.1 | 2 | 140 |

Genomic Map