Assay Details
Target Gene Details
Entrez Gene ID: | 67694 |
Gene Name: | intraflagellar transport 74 |
Gene Aliases: |
1700029H06Rik, Ccdc2, Cmg1, b2b796Clo |
Location: |
Chr.4:94603266-94693233 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift74 | NM_001290568.1 | NP_001277497.1 | ||
| NM_026319.3 | NP_080595.2 | |||
| AK006492.1 | BAB24617.1 | |||
| AK011690.1 | BAB27780.1 | |||
| AK053369.1 | BAC35365.1 | |||
| BC005798.1 | ||||
| BC023760.1 | AAH23760.1 | |||
| BY706530.1 |
More Information
Set Membership: |
|

Genomic Map