Assay Details
Target Gene Details
Entrez Gene ID: | 12757 |
Gene Name: | clathrin, light polypeptide (Lca) |
Gene Aliases: |
AV026556, Lca |
Location: |
Chr.4:44003816-44032848 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clta | NM_001080384.1 | NP_001073853.1 | ||
| NM_001080385.1 | NP_001073854.1 | |||
| NM_001080386.1 | NP_001073855.1 | |||
| NM_001290470.1 | NP_001277399.1 | |||
| NM_016760.2 | NP_058040.2 | |||
| AB575971.1 | ||||
| AK146965.1 | BAE27571.1 | |||
| AK149761.1 | BAE29067.1 | |||
| AK150148.1 | BAE29342.1 | |||
| AK152397.1 | BAE31185.1 | |||
| AK168130.1 | BAE40099.1 | |||
| AK216692.1 | ||||
| BC057660.1 | AAH57660.1 | |||
| BU847112.1 | ||||
| CA322069.1 | ||||
| CX243404.1 | ||||
| CX244279.1 | ||||
| U91848.1 | AAB51286.1 |
More Information
Set Membership: |
|

Genomic Map