Assay Details
Target Gene Details
Entrez Gene ID: | 623474 |
Gene Name: | RAD54 homolog B (S. cerevisiae) |
Gene Aliases: |
E130016E03Rik |
Location: |
Chr.4:11558920-11615808 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad54b | NM_001039556.3 | 3 | 303 | NP_001034645.1 |
| NM_001256145.1 | 3 | 303 | NP_001243074.1 | |
| AK078439.1 | 3 | 281 | BAC37274.1 | |
| BC057604.1 | 3 | 251 | AAH57604.1 | |
| CJ160401.1 | 3 | 304 |

Genomic Map