Assay Details
Target Gene Details
Entrez Gene ID: | 15356 |
Gene Name: | 3-hydroxy-3-methylglutaryl-Coenzyme A lyase |
Gene Aliases: |
AW476067, HL |
Location: |
Chr.4:135946448-135962617 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmgcl | NM_008254.2 | NP_032280.2 | ||
| AK077842.1 | ||||
| AK132035.1 | BAE20956.1 | |||
| AK144768.1 | BAE26056.1 | |||
| AK145251.1 | BAE26328.1 | |||
| AK154033.1 | BAE32329.1 | |||
| BB852743.1 | ||||
| BC025440.1 | AAH25440.1 | |||
| S65036.1 | AAB27965.1 |
More Information
Set Membership: |
|

Genomic Map