Assay Details
Target Gene Details
Entrez Gene ID: | 13370 |
Gene Name: | deiodinase, iodothyronine, type I |
Gene Aliases: |
5DI, D1, ITDI1, TXDI1 |
Location: |
Chr.4:107291465-107307143 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dio1 | NM_007860.3 | NP_031886.3 | ||
| AK002549.1 | ||||
| BC019786.1 | ||||
| BC037080.1 | AAH37080.1 | |||
| BC089608.1 | AAH89608.1 | |||
| U49861.1 | AAB00228.1 |

Genomic Map