Assay Details
Target Gene Details
Entrez Gene ID: | 230082 |
Gene Name: | nucleolar protein family 6 (RNA-associated) |
Gene Aliases: |
AA410091, Nrap |
Location: |
Chr.4:41114427-41124339 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol6 | NM_139236.3 | 25 | 3352 | NP_631982.2 |
| AF361077.1 | 25 | 3301 | AAL74401.1 | |
| AF361078.1 | AAL74402.1 | |||
| AK089138.1 | 25 | 3285 | ||
| AK155818.1 | 25 | 3291 | BAE33447.1 | |
| AK166559.1 | 25 | 3279 | ||
| AK166650.1 | 25 | 3278 | BAE38917.1 | |
| AK212631.1 | 1 | 54 | ||
| BC019981.1 | 11 | 1307 | AAH19981.1 | |
| BC021856.1 | 11 | 1357 | AAH21856.1 | |
| BC059820.1 | 25 | 3270 |
More Information
Set Membership: |
|

Genomic Map