Assay Details
Target Gene Details
Entrez Gene ID: | 230082 |
Gene Name: | nucleolar protein family 6 (RNA-associated) |
Gene Aliases: |
AA410091, Nrap |
Location: |
Chr.4:41114427-41124339 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nol6 | NM_139236.3 | 3 | 445 | NP_631982.2 |
| AF361077.1 | 3 | 394 | AAL74401.1 | |
| AF361078.1 | 3 | 394 | AAL74402.1 | |
| AK053117.1 | BAC35271.1 | |||
| AK089138.1 | 3 | 365 | ||
| AK155818.1 | 3 | 384 | BAE33447.1 | |
| AK166559.1 | 3 | 371 | ||
| AK166650.1 | 3 | 371 | BAE38917.1 | |
| AK171163.1 | 3 | 382 | BAE42285.1 | |
| AK171421.1 | 3 | 372 | BAE42443.1 | |
| AK191861.1 | ||||
| AK195706.1 | ||||
| AK197974.1 | ||||
| AK198310.1 | ||||
| AK199838.1 | ||||
| AK202668.1 | ||||
| AK204352.1 | ||||
| AK209500.1 | ||||
| BC059820.1 | 3 | 363 | ||
| CN459855.1 | 3 | 445 |
More Information
Set Membership: |
|

Genomic Map