Assay Details
Target Gene Details
Entrez Gene ID: | 59021 |
Gene Name: | RAB2A, member RAS oncogene family |
Gene Aliases: |
9330148M11Rik, C80220, Rab2 |
Location: |
Chr.4:8535586-8607702 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab2a | NM_021518.3 | NP_067493.1 | ||
| AB232585.1 | BAF02847.1 | |||
| AK042862.1 | BAC31385.1 | |||
| AK079071.1 | BAC37524.1 | |||
| AK135421.1 | BAE22525.1 | |||
| AK146179.1 | BAE26957.1 | |||
| AK150030.1 | BAE29254.1 | |||
| AK159607.1 | BAE35226.1 | |||
| AK160333.1 | BAE35742.1 | |||
| AK163320.1 | BAE37298.1 | |||
| AK184879.1 | ||||
| AK194278.1 | ||||
| BC058382.1 | AAH58382.1 | |||
| X95403.1 | CAA64684.1 |

Genomic Map