Assay Details
Target Gene Details
Entrez Gene ID: | 19359 |
Gene Name: | RAD23b homolog (S. cerevisiae) |
Gene Aliases: |
0610007D13Rik, AV001138, HR23B, mHR23B, p58 |
Location: |
Chr.4:55350042-55392237 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad23b | NM_009011.4 | NP_033037.2 | ||
| AK084917.1 | 2 | 1767 | ||
| AK142280.1 | BAE25006.1 | |||
| AK150089.1 | BAE29298.1 | |||
| AK157252.1 | BAE34014.1 | |||
| AK160880.1 | BAE36067.1 | |||
| AK160890.1 | BAE36071.1 | |||
| AK160973.1 | BAE36124.1 | |||
| AK167582.1 | BAE39643.1 | |||
| AK177667.1 | ||||
| AK191948.1 | ||||
| AK203724.1 | ||||
| AK210573.1 | ||||
| AK218197.1 | ||||
| BC027747.1 | AAH27747.1 | |||
| BC068193.1 | AAH68193.1 | |||
| X92411.1 | CAA63146.1 |
More Information
Set Membership: |
|

Genomic Map